Hypodermic Needles, Mortech
12, 127.0 mm (5.00")
76549-810
: Mopec
: FC912-22
Hypodermic needles are designed for aspiration/cavity injection or extraction.
- Stainless steel, long lasting piercing points, Luer hubs.
Available in a variety of sizes.
- Length:127.0 mm (5.00")
- Gauge:12