Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Hypodermic Needles, Mortech

Hypodermic Needles, Mortech

20, 63.5 mm (2.50")
Catalog # 76549-790
Supplier:   Mopec
Supplier #:  FC912-12
Hypodermic Needles, Mortech
Catalog # 76549-790
Supplier:  Mopec
Hypodermic Needles, Mortech
Hypodermic Needles, Mortech

20, 63.5 mm (2.50")

Price
$4.73
Log in for availability
Each
Product Details & Documents

Hypodermic needles are designed for aspiration/cavity injection or extraction.

  • Stainless steel, long lasting piercing points, Luer hubs.

Available in a variety of sizes.

Specifications
  • Length:
    63.5 mm (2.50")
  • Gauge:
    20