Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Platform Scale, Mortech

Standard platform scale, 2200 g, 140×170 mm (5.5×6.7 in.), 0,1 g
  76549-586
 :   Mopec
 :  BB065
Platform Scale, Mortech
  76549-586
 :  Mopec
Platform Scale, Mortech
Platform Scale, Mortech

Standard platform scale, 2200 g, 140×170 mm (5.5×6.7 in.), 0,1 g

 
$967.77
   
Each
 

Ideal for laboratory and industrial applications, our standard and premium Platform Scale features an easy to navigate color touchscreen display and a slim design allowing for stackable storage. Geared for superior performance with fast stabilization time and high resolution weighing results.

  • The Standard platform scale features multiple weighing units; USB host, USB device or RS232 connectivity; battery or AC power.
  • The Premium platform scale includes RS232 interface –allows data connection to PC or printer; AC adapter included; GLP/GMP data – capabilities including date, time, balance serial number, user ID/project ID, and results.

Features also include superior overload protection and expanded connectivity options. Available in standard or premium.

 
  • Pan/Platform Size:
    140×170 mm (5.5×6.7 in.)
  • Description:
    Standard platform scale
  • Readability:
    0,1 g
  • Weighing capacity:
    2200 g