Knife Sharpener, Variable Speed, Mortech
Product Details & Documents
This variable speed sharpener creates a precise angle and a consistent, exact bevel by sharpening, thinning, and honing which results in a stronger cutting edge. The honing wheels are mounted on parallel shafts that rotate in opposite directions.
- Stainless steel case
Available in US and International voltage.
Ordering information: Replacement honing wheels available (See model AH034).
Specifications
- Description:220 V