Orders LinkContactUsLinkComponent
Scissors, Enterotomy, Mortech
Scissors, Enterotomy, Mortech
Standard Grade, 8.25"
Catalog # 76549-288
Supplier:   Mopec
Supplier #:  AH003
Scissors, Enterotomy, Mortech
Catalog # 76549-288
Supplier:  Mopec
Scissors, Enterotomy, Mortech
Scissors, Enterotomy, Mortech
Standard Grade, 8.25"
Price
$99.39
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Sturdy, large, gastrointestinal scissors. Designed with the ability to pull away tissue.
  • Stainless steel
  • Hook blade tip
Available in standard or premium grade.
Specifications
  • Description:
    Standard Grade
  • Length:
    8.25"