Orders LinkContactUsLinkComponent
Forceps, Dissection, Toothed, Mortech
Standard, 10", 1×2
Catalog # 76548-652
Supplier:   Mopec
 :  AB015
Forceps, Dissection, Toothed, Mortech
Catalog # 76548-652
Supplier:  Mopec
Forceps, Dissection, Toothed, Mortech
Forceps, Dissection, Toothed, Mortech
Standard, 10", 1×2
Price
$44.09
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Post-mortem dissection tooth forceps are available in a wide variety of lengths and tooth patterns.
  • Stainless steel
  • Ribbed grip
  • Varied teeth standard pattern tips
Available in standard or premium grade with various teeth patterns.
Specifications
  • Length:
    10"
  • No. of teeth:
    1×2
  • Grade:
    Standard