Scissors, Disposable, Mortech
Product Details & Documents
These disposable scissors are highly recommended for post-mortem infectious cases.
- Straight cutting edge
- Not autoclavable
- Single use only
Package of 12. Available in multiple lengths and tips.
Specifications
- Length:5.25"
- Blade type:Sharp/Blunt