Orders LinkContactUsLinkComponent
Scissors, SuperCut, Sharp/Blunt, Mortech
Scissors, SuperCut, Sharp/Blunt, Mortech
5.5", Straight
Catalog # 76548-570
Supplier:   Mopec
Supplier #:  AA508
Scissors, SuperCut, Sharp/Blunt, Mortech
Catalog # 76548-570
Supplier:  Mopec
Scissors, SuperCut, Sharp/Blunt, Mortech
Scissors, SuperCut, Sharp/Blunt, Mortech
5.5", Straight
Price
$285.69
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
SuperCut scissors are tempered and hardened for an extended, sharper life over ordinary steel. The SuperCut blade has a specially designed sharpened edge. They also feature rounded and polished finger rings. A general tool for cutting.
  • Stainless steel
  • Sharp/blunt tips
Available with straight or curved edges.
Specifications
  • Length:
    5.5"
  • Curvature:
    Straight