Orders LinkContactUsLinkComponent
Scissors, Superfine Micro, Mortech
Scissors, Superfine Micro, Mortech
3.25", Straight
Catalog # 76548-492
Supplier:   Mopec
Supplier #:  AA152
Scissors, Superfine Micro, Mortech
Catalog # 76548-492
Supplier:  Mopec
Scissors, Superfine Micro, Mortech
Scissors, Superfine Micro, Mortech
3.25", Straight
Price
$749.88
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Delicate scissors with a fine point for small, very precise, post-mortem dissection.
  • Premium grade stainless steel
  • Superfine sharp tips
Available with straight or curved edges.
Specifications
  • Length:
    3.25"
  • Curvature:
    Straight