Orders LinkContactUsLinkComponent
Scissors, Iris, Curved, Mortech
Premium Grade, 4.5"
  76548-458
 :   Mopec
 :  AA129
Scissors, Iris, Curved, Mortech
  76548-458
 :  Mopec
Scissors, Iris, Curved, Mortech
Scissors, Iris, Curved, Mortech
Premium Grade, 4.5"
$116.74
Each
Ideal for small, precise, post-mortem dissection.
  • Stainless steel
  • Curve cutting edge
  • Sharp points
Available in standard or premium grade.
  • Description:
    Premium Grade
  • Length:
    4.5"