Scissors, Dissection, Sharp/Blunt, Straight, Mortech
Product Details & Documents
These scissors are designed with one sharp tip, one blunt tip and a straight cutting edge. Ideal for a variety of post-mortem dissection needs.
- Stainless steel
- Straight cutting edge
- Sharp/blunt tips
Available in standard or premium grade.
Specifications
- Description:Premium Grade
- Length:5.5"