Orders LinkContactUsLinkComponent
Mounting Media Acrymount™ Plus, Statlab
Mounting Media Acrymount™ Plus, Statlab
Mounting Media, Acrymount™ Plus, 4 oz
Catalog # 76548-258
Supplier:   StatLab
Supplier #:  SL80PLUS-4
Mounting Media Acrymount™ Plus, Statlab
Catalog # 76548-258
Supplier:  StatLab
CAS Number:  
Mounting Media Acrymount™ Plus, Statlab
Mounting Media Acrymount™ Plus, Statlab
Mounting Media, Acrymount™ Plus, 4 oz
CAS Number:  
Price
$56.42
Log in for availability
Pack of 4 (1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A rapid-drying, toluene-based mounting medium/liquid coverglass. Compatible with xylene, and xylene-substitute clearing agents including d-Limonene.
  • Slides mounted with Acrymount™ Plus may be screened in as little as 10 minutes.
  • Formulated to reduce oxidation, preventing yellowing and fading in H&E, IHC and special stains after coverslipping
  • Maintains an optimal refractive index for viewing and photo microscopy
  • For labs seeking to eliminate final clearing with xylene, tissue may be coverslipped directly from absolute Isopropyl Alcohol
  • Compatible with both manual and most automated coverslipping processes.
Part of the StatLab family of liquid mounting media.
Manufactured in the USA.
Documents
Specifications
Specifications
  • Description:
    Mounting Media, Acrymount™ Plus
  • Size:
    4 oz