Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Mounting Media Acrymount™ Plus, Statlab

Mounting Media Acrymount™ Plus, Statlab

Mounting Media, Acrymount™ Plus, 16 oz
Catalog # 76548-256
Supplier:   StatLab
Supplier #:  SL80PLUS-16
Mounting Media Acrymount™ Plus, Statlab
Catalog # 76548-256
Supplier:  StatLab
CAS Number:  
Mounting Media Acrymount™ Plus, Statlab
Mounting Media Acrymount™ Plus, Statlab

Mounting Media, Acrymount™ Plus, 16 oz

CAS Number:  
Price
$52.70
Log in for availability
Each (16oz)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

A rapid-drying, toluene-based mounting medium/liquid coverglass. Compatible with xylene, and xylene-substitute clearing agents including d-Limonene.

  • Slides mounted with Acrymount™ Plus may be screened in as little as 10 minutes.
  • Formulated to reduce oxidation, preventing yellowing and fading in H&E, IHC and special stains after coverslipping
  • Maintains an optimal refractive index for viewing and photo microscopy
  • For labs seeking to eliminate final clearing with xylene, tissue may be coverslipped directly from absolute Isopropyl Alcohol
  • Compatible with both manual and most automated coverslipping processes.

Part of the StatLab family of liquid mounting media.

Manufactured in the USA.

Documents

Specifications

Specifications

  • Description:
    Mounting Media, Acrymount™ Plus
  • Size:
    16 oz

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.