Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Tissue Marking Dyes, StatLab

Tissue Marking Dyes, StatLab

Tissue Marking Dye Set, Assorted colours, 1 case
Catalog # 76548-254
Supplier:   StatLab
Supplier #:  SL669
Tissue Marking Dyes, StatLab
Catalog # 76548-254
Supplier:  StatLab
CAS Number:  
Tissue Marking Dyes, StatLab
Tissue Marking Dyes, StatLab

Tissue Marking Dye Set, Assorted colours, 1 case

CAS Number:  
Price
$332.46
Log in for availability
Case (2oz)
Product Details & Documents

Permanent tissue marking dyes. Seven colors formulated to define margins without bleeding or fading. Durable, bright dyes hold permanently through processing and staining procedures. Unique lids may either be completely removed or conveniently flipped open to prevent drying.

Set includes one tissue marking dye rack and a set of 2 oz dyes in each of 7 colors.

Packaging: Manufactured in the USA

Specifications

Specifications

  • Color:
    Assorted colours
  • Description:
    Tissue Marking Dye Set
  • Size:
    1 case

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.