Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Prostate Biopsy Kit, StatLab

Prostate Biopsy Kit, StatLab

Prostate biopsy kit, 20 ml (12 Vials per kit)
Catalog # 76548-180
Supplier:   StatLab
Supplier #:  PBKNB12
Prostate Biopsy Kit, StatLab
Catalog # 76548-180
Supplier:  StatLab
Prostate Biopsy Kit, StatLab
Prostate Biopsy Kit, StatLab

Prostate biopsy kit, 20 ml (12 Vials per kit)

Price
$24.98
Log in for availability
Each (20ml)
Product Details & Documents

All StatLab prostate biopsy kits are pre-labeled with a location diagram supporting accurate collection. Kits contain prefilled StatClick™ vials with 10% neutral buffered formalin solution arranged in pre-labeled positions that correlate with the area of the prostate from which sample is taken, and stickers to affix to the prefill indicating the biopsy location. 6, 8, 12, or 16-vial biopsy kits are available. StatClick patented vials produce an audible “clicking” sound when the leak-proof seal has been properly engaged, assuring accurate closure and maintaining specimen integrity during transport.

Packaging: Manufactured in the USA

Specifications
  • Description:
    Prostate biopsy kit
  • Size:
    20 ml (12 Vials per kit)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.