Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Formalin 10% Modified Carson/Millonig

Formalin 10% Modified Carson/Millonig

20 ml (96 per case)
Catalog # 76548-098
Corrosive
Irritant
Health
Supplier:   StatLab
Supplier #:  CM1507
Formalin 10% Modified Carson/Millonig
Catalog # 76548-098
Supplier:  StatLab
CAS Number:  50-00-0
Formalin 10% Modified Carson/Millonig
Formalin 10% Modified Carson/Millonig

20 ml (96 per case)

CAS Number:  50-00-0
Price
$72.20
Log in for availability
Case of 96 (20ml)
Product Details & Documents

A modified buffering formulation to preserve tissue.

  • Carson Millonig modified buffering formulation preserves tissue specimens
  • For both light and electron microscopy
  • Can be used in place of traditional 10% NBF

Part of the StatLab line of StatClick 10% NBF Prefills.

Manufactured in the USA

Synonyms:  Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
Specifications

Specifications

  • Size:
    20 ml (96 per case)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.