Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Adenosine 99+%

Adenosine 99+%

Glass bottle, 100 g
Catalog # TS16404-1000
Supplier #:  16404-1000
Adenosine 99+%
Catalog # TS16404-1000
CAS Number:  58-61-7
Adenosine 99+%
Adenosine 99+%

Glass bottle, 100 g

CAS Number:  58-61-7
Price
$321.00
Log in for availability
Each (100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
  • CAS Number:58-61-7
  • Formula:C₁₀H₁₃N₅O₄
  • MDL Number:MFCD00005752
  • Molecular Weight:267.24 g/mol
  • Melting Pt:233…238 °C
  • Storage Temperature:Ambient
  • EINECS:200-389-9

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    233 to 238 °C
  • Loss on drying:
    ≤0.5 % (105 °C, 2 hrs)
  • Sulfated ash:
    ≤0.1 %
  • HPLC:
    ≥99.0 %
  • UV spectrophotometry:
    98.0 to 102.0 % (lambda max. 259 nm, pH 7)
  • UV transmittance:
    ≥98 % (c=0.5, H2O, 430 nm) 1 cm cell
  • Ratio of Absorbances:
    0.76 to 0.80 (A250/A260)
  • Nitrogen (N):
    25.69 to 26.73 % (on dry basis)
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.