D-(-)-Sorbitol 97%
Plastic bottle, 2.5 kg
Catalog # TS13273-0025
Supplier: Thermo Scientific Chemicals
: 13273-0025
Jump to:
Product Details & Documents
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
- CAS Number:50-70-4
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00004708
- Molecular Weight:182.17 g/mol
- Merck Index:13,08797
- Melting Pt:95…99 °C
- Storage Temperature:Ambient
- EINECS:200-061-5
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤1.5 %
- Sulfated ash:≤0.1 %
- HPLC:≥96.0 % (on anhydrous substance)
- Specific optical rotation:+4.0° to +7.0° (20°C, 589 nm) (c=10, borate soln.) (on anhydrous substance)
- Lead (Pb):≤0.5 ppm
- Nickel (Ni):≤1 ppm
Specifications
- Pack type:Plastic bottle
- Size:2.5 kg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



