D-(-)-N-Methylglucamine 99%
Plastic bottle, 500 g
Catalog # TS12684-5000
Supplier: Thermo Scientific Chemicals
: 12684-5000
D-(-)-N-Methylglucamine 99%
Catalog # TS12684-5000
Supplier: Thermo Scientific Chemicals
CAS Number: 6284-40-8
Jump to:
Product Details & Documents
Synonyms: Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol
- CAS Number:6284-40-8
- Formula:H₃CNHCH₂(CH(OH))₄CH₂OH
- MDL Number:MFCD00004707
- Molecular Weight:195.22 g/mol
- Merck Index:12,06154
- Melting Pt:128…132 °C
- Storage Temperature:Ambient
- EINECS:228-506-9
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:128 to 132 °C
- Loss on drying:≤1 % (1 g, 100 °C, 3 hrs)
- Sulfated ash:≤0.1 %
- UV:(20 % w/v in water)
- Specific optical rotation:-15.7° to -17.3° (20 °C, 589 nm) (c=10, H2O) (on dried substance)
- Titration with HClO4:≥98.5 % (On dried substance)
- Chloride (Cl):≤100 ppm
- Sulfate (SO4):≤150 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤20 ppm
Specifications
- Pack type:Plastic bottle
- Size:500 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



