Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Formalin 10% Modified Carson/Millonig

Formalin 10% Modified Carson/Millonig

60 mL (96 per case)
Catalog # 76548-094
Corrosive
Irritant
Health
Supplier:   StatLab
Supplier #:  CM1230
Formalin 10% Modified Carson/Millonig
Catalog # 76548-094
Supplier:  StatLab
CAS Number:  50-00-0
Formalin 10% Modified Carson/Millonig
Formalin 10% Modified Carson/Millonig

60 mL (96 per case)

CAS Number:  50-00-0
Price
$101.20
Log in for availability
Case of 96 (60ml)
Product Details & Documents

A modified buffering formulation to preserve tissue.

  • Carson Millonig modified buffering formulation preserves tissue specimens
  • For both light and electron microscopy
  • Can be used in place of traditional 10% NBF

Part of the StatLab line of StatClick 10% NBF Prefills.

Manufactured in the USA

Synonyms:  Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
Specifications

Specifications

  • Size:
    60 mL (96 per case)

Specifications