Orders LinkContactUsLinkComponent
Fixative Dissect Aid™, Statlab
Fixative Dissect Aid™, Statlab
Fixative Dissect Aid™, Cube, 5 gallon
Catalog # 76548-066
Supplier:   StatLab
Supplier #:  9001-5
Fixative Dissect Aid™, Statlab
Catalog # 76548-066
Supplier:  StatLab
CAS Number:  
Fixative Dissect Aid™, Statlab
Fixative Dissect Aid™, Statlab
Fixative Dissect Aid™, Cube, 5 gallon
CAS Number:  
Price
$224.23
Log in for availability
Each (5US Gal)
Product Details & Documents
Dissect-aid is a fixative which helps make lymph nodes more identifiable during pathology examination.
  • The product turns lymph nodes pale/white so they stand out from other tissue
  • Solution firms the nodes for touch identification when performing gross examination
  • Dramatically improves lymph node recovery/harvesting, and also an excellent fixative for fatty tissues
  • This formulation has helped in diagnosis for the past 30 years
Part of the StatLab line of prefilled bulk containers.
Manufactured in the USA
Specifications
Specifications
  • Pack type:
    Cube
  • Description:
    Fixative Dissect Aid™
  • Size:
    5 gallon