Orders LinkContactUsLinkComponent
Fixative Dissect Aid™, Statlab
Fixative Dissect Aid™, 1 gallon
  76548-062
 :  9001-1
Fixative Dissect Aid™, Statlab
  76548-062
 :  
 :  
Fixative Dissect Aid™, Statlab
Fixative Dissect Aid™, Statlab
Fixative Dissect Aid™, 1 gallon
 :  
Dissect-aid is a fixative which helps make lymph nodes more identifiable during pathology examination.
  • The product turns lymph nodes pale/white so they stand out from other tissue
  • Solution firms the nodes for touch identification when performing gross examination
  • Dramatically improves lymph node recovery/harvesting, and also an excellent fixative for fatty tissues
  • This formulation has helped in diagnosis for the past 30 years
Part of the StatLab line of prefilled bulk containers.
Manufactured in the USA
  • Description:
    Fixative Dissect Aid™
  • Size:
    1 gallon