Formaldehyde 37%
Cube, 5 gallon
Catalog # 76548-026
Supplier: StatLab
Supplier #: 28530-5
Jump to:
Product Details & Documents
37% Formaldehyde solution.
- Contains methanol as a stabilizer
Part of the StatLab line of prefilled bulk containers
Manufactured in the USA
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- UN:2209
- ADR:8,III
Specifications
Specifications
- Pack type:Cube
- Size:5 gallon
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



