Orders LinkContactUsLinkComponent
Formaldehyde 37%
1 gallon
  76548-024
 :  28530-1
Formaldehyde 37%
  76548-024
 :  
 :  50-00-0
Formaldehyde 37%
Formaldehyde 37%
1 gallon
 :  50-00-0
37% Formaldehyde solution.
  • Contains methanol as a stabilizer
Part of the StatLab line of prefilled bulk containers
Manufactured in the USA
 :  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  •  :50-00-0
  •  :CH₂O
  •  :MFCD00003274
  •  :30.03 g/mol
  •  :2209
  •  :8,III
  • Size:
    1 gallon