Orders LinkContactUsLinkComponent
Formical-2000™ Decalcifier, StatLab
Formical-2000™ Decalcifier, 5 Gallon (Cube)
Catalog # 76548-018
Supplier:   StatLab
 :  1314-5
Formical-2000™ Decalcifier, StatLab
Catalog # 76548-018
Supplier:  StatLab
CAS Number:  
Formical-2000™ Decalcifier, StatLab
Formical-2000™ Decalcifier, StatLab
Formical-2000™ Decalcifier, 5 Gallon (Cube)
CAS Number:  
Price
$202.80
Log in for availability
Each (5US Gal)
Product Details & Documents
A fast yet gentle bone decalcifier that has been formulated from semiconductor grade formic acid, highly purified reagent grade water and EDTA. Formical-2000™ works well not only as a general purpose decalcifier for all routine histological specimens, it works especially well to decalcify tissue for immunohistochemistry. Will safely decalcify bone without destroying antigen sites.
Packaging: Manufactured in the USA
Specifications
Specifications
  • Description:
    Formical-2000™ Decalcifier
  • Size:
    5 Gallon (Cube)