Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Formical-2000™ Decalcifier, StatLab

Formical-2000™ Decalcifier, StatLab

Formical-2000™ Decalcifier, 32 oz
Catalog # 76548-016
Supplier:   StatLab
Supplier #:  1314-32
Formical-2000™ Decalcifier, StatLab
Catalog # 76548-016
Supplier:  StatLab
CAS Number:  
Formical-2000™ Decalcifier, StatLab
Formical-2000™ Decalcifier, StatLab

Formical-2000™ Decalcifier, 32 oz

CAS Number:  
Price
$35.67
Log in for availability
Each (32oz)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

A fast yet gentle bone decalcifier that has been formulated from semiconductor grade formic acid, highly purified reagent grade water and EDTA. Formical-2000™ works well not only as a general purpose decalcifier for all routine histological specimens, it works especially well to decalcify tissue for immunohistochemistry. Will safely decalcify bone without destroying antigen sites.

Packaging: Manufactured in the USA

Specifications

Specifications

  • Description:
    Formical-2000™ Decalcifier
  • Size:
    32 oz

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.