Orders LinkContactUsLinkComponent
Formical-2000™ Decalcifier, StatLab
Formical-2000™ Decalcifier, StatLab
Formical-2000™ Decalcifier, 1 Gallon
Catalog # 76548-014
Supplier #:  1314-1
Formical-2000™ Decalcifier, StatLab
Catalog # 76548-014
Supplier:  
CAS Number:  
Formical-2000™ Decalcifier, StatLab
Formical-2000™ Decalcifier, StatLab
Formical-2000™ Decalcifier, 1 Gallon
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
DECALCIFIER FORMICAL-2000 32 OZ Catalog #76548-016
Product Details & Documents
A fast yet gentle bone decalcifier that has been formulated from semiconductor grade formic acid, highly purified reagent grade water and EDTA. Formical-2000™ works well not only as a general purpose decalcifier for all routine histological specimens, it works especially well to decalcify tissue for immunohistochemistry. Will safely decalcify bone without destroying antigen sites.
 : Manufactured in the USA
Specifications
Specifications
  • Description:
    Formical-2000™ Decalcifier
  • Size:
    1 Gallon
  • Cat. No.:
    76548-014
Specifications