Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Fixative Bouins, Statlab

Fixative Bouins, Statlab

Bouins Fixative, 32 oz
Catalog # 76547-978
Supplier:   StatLab
Supplier #:  11750-32
Fixative Bouins, Statlab
Catalog # 76547-978
Supplier:  StatLab
CAS Number:  
Fixative Bouins, Statlab
Fixative Bouins, Statlab

Bouins Fixative, 32 oz

CAS Number:  
Price
$39.05
Log in for availability
Each (32oz)
Product Details & Documents

Saturated aqueous picric acid/formalin solution.

  • Bouins is commonly used in trichrome staining procedures and with GI tract specimens

Packaging: Manufactured in the USA

Specifications

Specifications

  • Description:
    Bouins Fixative
  • Size:
    32 oz

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.