Orders LinkContactUsLinkComponent
Fixative Bouins, Statlab
Bouins Fixative, 60 ml (96 per case)
  76547-976
 :   StatLab
 :  11750-2
Fixative Bouins, Statlab
  76547-976
 :  StatLab
 :  
Fixative Bouins, Statlab
Fixative Bouins, Statlab
Bouins Fixative, 60 ml (96 per case)
 :  
$159.81
Case of 96 (60ml)
Saturated aqueous picric acid/formalin solution.
  • Bouins is commonly used in trichrome staining procedures and with GI tract specimens
 : Manufactured in the USA
  • Description:
    Bouins Fixative
  • Size:
    60 ml (96 per case)