Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Fixative Bouins, Statlab

Fixative Bouins, Statlab

Bouins Fixative, 40 ml (96 per case)
Catalog # 76547-974
Supplier:   StatLab
Supplier #:  11750-1
Fixative Bouins, Statlab
Catalog # 76547-974
Supplier:  StatLab
CAS Number:  
Fixative Bouins, Statlab
Fixative Bouins, Statlab

Bouins Fixative, 40 ml (96 per case)

CAS Number:  
Price
$118.52
Log in for availability
Case of 96 (40ml)
Product Details & Documents

Saturated aqueous picric acid/formalin solution.

  • Bouins is commonly used in trichrome staining procedures and with GI tract specimens

Packaging: Manufactured in the USA

Specifications

Specifications

  • Description:
    Bouins Fixative
  • Size:
    40 ml (96 per case)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.