Orders LinkContactUsLinkComponent
Fixative Bouins, Statlab
Bouins Fixative, 20 ml (96 per case)
  76547-972
 :   StatLab
 :  11750-.5
Fixative Bouins, Statlab
  76547-972
 :  StatLab
 :  
Fixative Bouins, Statlab
Fixative Bouins, Statlab
Bouins Fixative, 20 ml (96 per case)
 :  
$99.15
Case of 96 (20ml)
Saturated aqueous picric acid/formalin solution.
  • Bouins is commonly used in trichrome staining procedures and with GI tract specimens
 : Manufactured in the USA
  • Description:
    Bouins Fixative
  • Size:
    20 ml (96 per case)