Hydrogen Peroxide Based Detergency Booster
1 x 330 gal., Tote
Catalog # 76547-758
Supplier: DIVERSEY INC.
Supplier #: 57782330
Jump to:
Product Details & Documents
Booster solution is a highly concentrated liquid additive that enhances detergency and soil removal where organic soiling is a problem.
- Enhances detergency and soil removal
- Removes organic deposits
- Improved detergency results in shorter cleaning time
- Improved detergency results in shorter cleaning time
When added to blended caustic detergents, Booster solution results in improved detergency over the use of blended products alone resulting in shorter cleaning times.
This product is suitable for automatic dosing by linking the additive pump to the caustic delivery pump, ensuring consistent delivery of product.
Care has to be taken to ensure that the correct level of product is dosed into the CIP system and that the oxygen generated is vented to air.
Documents
Specifications
- Size:1 x 330 gal.
- Packaging:Tote