Orders LinkContactUsLinkComponent
Vee Gee Snap Cap Replacements
#1, 5 and 10 ml flasks (76546-110)
Catalog # 76546-716
Supplier:   Heathrow Scientific
 :  SC#1
Vee Gee Snap Cap Replacements
Catalog # 76546-716
Vee Gee Snap Cap Replacements
Vee Gee Snap Cap Replacements
#1, 5 and 10 ml flasks (76546-110)
Price
$2.71
Log in for availability
Each
Product Details & Documents
Replacement snap-caps.
  • For SIBATA volumetric flasks
Sold by the each.
Specifications
  • Cap Size:
    #1
  • Fits:
    5 and 10 ml flasks (76546-110)