Baume Hydrometers
29...41° Baume, 305 mm (12"), 0.1° Baume
76546-464
: 6609-4
VEE GEE scientific Baume scale hydrometers are for liquids heavier than water. The Baume scale is based on the following: Specific gravity = 145/(145 - degrees Baume).
- Wide variety of Baume ranges available
Calibrated for use at 15.5 °C/60 °F.
- Length:305 mm (12")
- Range:29...41° Baume
- Subdivisions:0.1° Baume



