Precision Short Form Specific Gravity Hydrometers
0.940...1.010, 165 mm (6.5"), 0.001
Catalog # 76546-354
Supplier: Heathrow Scientific
Supplier #: 6602-6S
Precision Short Form Specific Gravity Hydrometers
Catalog # 76546-354
Supplier: Heathrow Scientific
Jump to:
Product Details & Documents
These precision short form specific gravity hydrometers use less volume of the test liquid while still offering 0.001 resolution.
Each of these hydrometers has a small specific gravity range and has been calibrated for use at 60 °F.
Specifications
- Length:165 mm (6.5")
- Range:0.940...1.010
- Subdivisions:0.001



