InfinityLab PlusPore, Agilent Technologies
Polypore 250×46 mm, Analytical, PolyPore, 5 µm, 4.60 mm, 250 mm, 200 - 2000000 Da, Mixed Pore Size
Catalog # AGPL1513-5500
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Supplier #: PL1513-5500
InfinityLab PlusPore, Agilent Technologies
Catalog # AGPL1513-5500
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Jump to:
Product Details & Documents
PlusPore GPC columns offer the highest resolution gel permeation chromatography.
- PlusPore GPC columns offer a major resolution improvement over traditional mixed medias and single pore columns
- High-efficiency small particles use multipore technology, maximizing resolution without sacrificing molecular weight range or lifetime
- OligoPore uses unique ultrahigh pore volume media to achieve superior separation of polymers below 3,300 Daltons
PLgel individual pore size GPC columns offer the highest resolution within a narrow molecular weight range. Columns may be stacked to extend the molecular weight range. Individual pore columns are generally recommended for very small and very large polymers, which can be difficult to resolve using more common PLgel MIXED columns.
Specifications
- Column Format:Analytical
- Ø int.:4.60 mm
- Particle Size:5 µm
- Description:Polypore 250×46 mm
- Length:250 mm
- Packing material:Mixed Pore Size
- Phase:PolyPore
- Exclusion limit:200 - 2000000 Da
- Cat. No.:AGPL1513-5500
- pH Range:1-14
- Temperature Limit:150 °C
- Max. Pressure:150 bar
- Wetted Materials:Ethylbenzene
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



