CrossLab Flow Cells for HPLC, Agilent Technologies
Capillary stainless steel for flow cells 15 - 40 ml/min flow rates, preparative compatibility, SI (Swagelok, 1.6 mm Int.Ø, stainless steel, intermediate screw), —, 0.5 mm, —
Catalog # AG5500-1343
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
: 5500-1343
CrossLab Flow Cells for HPLC, Agilent Technologies
Catalog # AG5500-1343
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Jump to:
Product Details & Documents
High-quality flow cells for Agilent HPLC detectors to allow the best performance in all applications.
- Designed for highest chromatographic performance
- Flow cells for nano flow applications
Agilent provides a comprehensive range of high-quality flow cells for Agilent HPLC detectors to allow the best performance in all applications. Available solutions address the requirements of protein research applications or assays to assess the integrity of bio-therapeutic compounds. Bio-inert flow cells are available for Agilent multiple wavelength and diode array UV-absorption detectors, as well as for special detection techniques such as fluorescence and dual-angle light scattering.
Specifications
- Volume:—
- Description:Capillary stainless steel for flow cells 15 - 40 ml/min flow rates, preparative compatibility, SI (Swagelok, 1.6 mm Int.Ø, stainless steel, intermediate screw)
- Pathlength:—
- Internal dimensions:0.5 mm