Orders LinkContactUsLinkComponent
Special Function Valves for HPLC, Agilent Technologies
Special Function Valves for HPLC, Agilent Technologies
InfinityLab active solvent modulation valve, for 2D-LC , 1300 bar, stainless steel
Catalog # AG5067-4266
Supplier #:  5067-4266
Special Function Valves for HPLC, Agilent Technologies
Catalog # AG5067-4266
Special Function Valves for HPLC, Agilent Technologies
Special Function Valves for HPLC, Agilent Technologies
InfinityLab active solvent modulation valve, for 2D-LC , 1300 bar, stainless steel
Price
$15,160.00
Log in for availability
Each
Product Details & Documents
High-quality supplies for special function valves such as 2D-LC valves.
  • Supplies for 2D-LC valve heads;2D-LC active flow modulation valves and supplies
  • Stream selector valves and supplies
The Agilent InfinityLab 2D-LC solution brings the separation power of two-dimensional LC (2D-LC) to every lab. This solution makes it easy to switch between one-dimensional UHPLC and all modes of 2D-LC, allowing you to adjust separation performance to your sample complexity, as well as guaranteeing highest usage of a single system.
Easy-to-use software enables fast method setup for heart-cutting 2D-LC, comprehensive 2D-LC, multiple heart-cutting 2D-LC, or Agilent's unique high-resolution sampling 2D-LC.
High-quality supplies, including rotor seals, stator heads and bearing rings, fit perfectly to your 2D-LC valves, ensuring optimal performance and minimal system downtime.
Specifications
  • Description:
    InfinityLab active solvent modulation valve, for 2D-LC , 1300 bar, stainless steel