Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Basic fuchsin 88%, pure

Basic fuchsin 88%, pure

Plastic drum, 2.5 kg
Catalog # TS41914-0025
Irritant
Health
Supplier #:  41914-0025
Basic fuchsin 88%, pure
Catalog # TS41914-0025
CAS Number:  632-99-5
Basic fuchsin 88%, pure
Basic fuchsin 88%, pure

Plastic drum, 2.5 kg

CAS Number:  632-99-5
Price
$1,030.00
Log in for availability
Each (2,500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
  • CAS Number:632-99-5
  • Formula:C₂₀H₂₀ClN₃
  • MDL Number:MFCD00012569
  • Molecular Weight:337.85 g/mol
  • Merck Index:12,05686
  • Melting Pt:250 °C (dec.)
  • Flash Pt:200 °C
  • Storage Temperature:Ambient
  • EINECS:211-189-6
Specifications

Specification Test Results

  • Appearance:
    Green Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤10 % (105°C)
  • Dye content:
    ≥83 % (on dry substance)
  • UV Lambda max:
    549 to 555 nm (in 50:50 IMS:water)
  • Absorption Ratio P-15/P+15:
    1.1 to 1.4
  • Specific absorbance A (1%/1cm):
    ≥1800 (at lambda max.)

Specifications

  • Pack type:
    Plastic drum
  • Size:
    2.5 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.