Vanillin 99%, pure
Product AAA11169-0E is similar to TS14082-2500 but may have a different manufacturer and/or packaging.
- :121-33-5
- :C₈H₈O₃
- :MFCD00006942
- :152.15 g/mol
- :13,09998
- :170 °C (15 mmHg)
- :81…84 °C
- :147 °C
- :Ambient
- :204-465-2
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:81 to 84 °C
- Titration with NaOH:≥98.5 %
- Pack type:Plastic bottle
- Size:250 g
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



