Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Vanillin 99%, pure

Vanillin 99%, pure

Glass bottle for solids, 2 g
Catalog # TS14082-0020
Irritant
Supplier #:  14082-0020
Vanillin 99%, pure
Catalog # TS14082-0020
CAS Number:  121-33-5
Vanillin 99%, pure
Vanillin 99%, pure

Glass bottle for solids, 2 g

CAS Number:  121-33-5
Price
$46.50
Log in for availability
Each (2g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,09998
  • Boiling Pt:170 °C (15 mmHg)
  • Melting Pt:81…84 °C
  • Flash Pt:147 °C
  • Storage Temperature:Ambient
  • EINECS:204-465-2
Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    81 to 84 °C
  • Titration with NaOH:
    ≥98.5 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Size:
    2 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.