Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
LED MOUNTED 6500 K 930 MW 1300MA 4.51V
Catalog # MSPP-MCWHL7
Supplier:
THORLABS INC MS
Expand image
LED MOUNTED 6500 K 930 MW 1300MA 4.51V
Catalog # MSPP-MCWHL7
Supplier:
THORLABS INC MS
Supplier Number:
MCWHL7