HUMAN CELL A549 EPITHELIAL LUNG FROZEN
Catalog # MSPP-CRMCCL185
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
HUMAN CELL A549 EPITHELIAL LUNG FROZEN
Catalog # MSPP-CRMCCL185
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
Supplier Number: CRM-CCL-185
Frequently Bought Together
Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade
Supplier: VWR
Pipet-Aid® XP² Ergonomic Pipetting Device, Drummond Scientific
Supplier: Drummond
Amersham ECL Western Blotting Select Detection Reagent, Cytiva
Supplier: Cytiva
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Supplier: Zymo Research
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemical
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
Radiance Plus Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
DNA BINDING BUFFER 100 ML
Supplier: Zymo Research
Anti-LY6K Mouse Monoclonal Antibody [clone: 750747]
Supplier: R&D Systems
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
PEROXIDE
Supplier: PROTEIN SIMPLE MS
Sodium dodecyl sulfate (SDS) solution 10% (w/v), Molecular Biology Grade
Supplier: Promega Corporation
FlashGel™ DNA, RNA and Recovery Systems
Supplier: Lonza
Human Recombinant IL4 (from <i>E. coli</i>)
Supplier: Peprotech
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: AB SCIEX LLC MS
Laemmli SDS sample buffer, reducing (6X), Thermo Scientific
Supplier: Thermo Scientific Chemicals