Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Copper etchant CE-100 semiconductor grade

HDPE bottle, Gallon
Catalog # 76541-002
Corrosive
Irritant
Health
Supplier:   Transene
 :  006CE100-GAL
Copper etchant CE-100 semiconductor grade
Catalog # 76541-002
Supplier:  Transene
CAS Number:  
Copper etchant CE-100 semiconductor grade

HDPE bottle, Gallon

CAS Number:  
Price
$147.67
Log in for availability
Each (1US Gal) Case of 4 (1 US Gal)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Copper Etchant CE-100 is a fast etch for removal of thicker copper films.

Synonyms:  Copper etching solution
  • MDL Number:MFCD00161912
  • Density:1.3 g/cc
  • Storage Temperature:Ambient
  • UN:2582
  • ADR:8,III
Specifications

Specification Test Results

  • Appearance:
    Brown
  • Etch Capacity (rate declines at ~70%):
    19 oz/gallon
  • Etch Rate at 40°C:
    1 (etches 1oz Cu in 90 sec) mL/minute
  • Filtration:
    1 um
  • Incompatible Materials:
    Al, Ni, Ti
  • pH:
    Acidic
  • Recommended Operating Temperatures:
    20-80°C (30-40°C most common) °C
  • Shelf Life:
    1 Year
  • Storage Conditions:
    Ambient

Specifications

  • Pack type:
    HDPE bottle
  • Size:
    Gallon

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.