Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Copper etchant APS-100 semiconductor grade

Copper etchant APS-100 semiconductor grade

HDPE bottle, Gallon
Catalog # 76540-926
Oxidizing
Irritant
Supplier:   Transene
Supplier #:  APS-100-GAL
Copper etchant APS-100 semiconductor grade
Catalog # 76540-926
Supplier:  Transene
CAS Number:  
Copper etchant APS-100 semiconductor grade

HDPE bottle, Gallon

CAS Number:  
Price
$118.56
Log in for availability
Each (1US Gal) Case of 4 (1 US Gal)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Copper Etchant APS-100 is an ammonium persulfate solution with excellent control for fine-line etching of copper films in integrated circuits.

Synonyms:  Copper etching solution
  • MDL Number:MFCD00161912
  • Boiling Pt:100 °C (water)
  • Density:1.08 g/cc
  • UN:3139
  • ADR:5.1,III
Specifications

Specification Test Results

  • Appearance:
    White-white
  • Etch Capacity (rate declines at ~70%):
    10 oz/gallon
  • Etch Rate at 40°C:
    80 (etches 1oz Cu in 7.5 min) Å/sec
  • Filtration:
    1 um
  • Incompatible Materials:
    Al, Ni
  • pH:
    Acidic
  • Recommended Operating Temperatures:
    20-80°C (30-40°C most common) °C
  • Shelf Life:
    1 Year
  • Storage Conditions:
    Ambient

Specifications

  • Pack type:
    HDPE bottle
  • Size:
    Gallon

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.