Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Lead(II) acetate basic ACS

Lead(II) acetate basic ACS

Plastic bottle, 5 kg
Catalog # TS45322-0050
Irritant
Health
Environment
Supplier #:  AC45322-0050
Lead(II) acetate basic ACS
Catalog # TS45322-0050
CAS Number:  1335-32-6
Lead(II) acetate basic ACS
Lead(II) acetate basic ACS

Plastic bottle, 5 kg

CAS Number:  1335-32-6
Price
$278.00
Log in for availability
Each (5kg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Horne′s compound, Lead(II) hydroxide acetate, Acetic acid lead (II) salt basic, Lead subacetate
  • CAS Number:1335-32-6
  • Formula:(CH₃COO)₂Pb·2Pb(OH)₂
  • MDL Number:MFCD00011154
  • Molecular Weight:807.72 g/mol
  • Storage Temperature:Ambient
  • UN:2291
  • ADR:6.1,III
Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Loss on drying:
    ≤1.5 % (105°C, 2 h)
  • Titration:
    ≥33.0 % Basic lead (PbO)
  • Chloride (Cl):
    ≤0.003 %
  • Nitrate (NO3):
    ≤0.003 %
  • Calcium (Ca):
    ≤0.01 %
  • Copper (Cu):
    ≤0.002 %
  • Iron (Fe):
    ≤0.002 %
  • Potassium (K):
    ≤0.02 %
  • Sodium (Na):
    ≤0.05 %
  • Insoluble matter:
    ≤0.02 % (in dilute acetic acid)
  • Insoluble matter in water:
    ≤1.0 %

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    5 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.