Universal® Wirebound Notebook, Medium/College Rule
Wirebound notebook, medium/college rule, Black, 152 mm (6"), 241 mm (9.5")
76540-152
: UNV66410
A clean and portable, wired notebook for virtually any note taking needs.
- Microperforated sheets provide a clean, easy tear
- Ruled lines encourage neat writing
- Durable wire spiral coil binding twist looped to resist snags, yet easy to open and flip
- Grayboard back cover material
Constructed with micro-perforated sheets which provide a clean, easy tear every time. The ruled lines encourage neat writing and make it easy to develop and outline or note structure. Wrapped with a durable wire coil binding that’s twist looped to resist snags. The wirebound design is easy to open and flip to write flat or folded.
- Cover color:Black
- Description:Wirebound notebook, medium/college rule
- Length:241 mm (9.5")
- Width:152 mm (6")



