Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Bismuth(III) hydroxide

Bismuth(III) hydroxide

Glass bottle, 250 g
Catalog # TS39329-2500
Supplier #:  AC39329-2500
Bismuth(III) hydroxide
Catalog # TS39329-2500
CAS Number:  10361-43-0
Bismuth(III) hydroxide
Bismuth(III) hydroxide

Glass bottle, 250 g

CAS Number:  10361-43-0
Price
$147.00
Log in for availability
Each (250g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Bismuth hydroxide
  • CAS Number:10361-43-0
  • Formula:H₃BiO₃
  • MDL Number:MFCD00046188
  • Molecular Weight:260
  • Melting Pt:415 °C
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Appearance:
    White to cream Powder
  • Loss on drying:
    ≤1.0 %
  • Identification:
    Positive
  • Bismuth (Bi):
    ≥80.0 % (on dry basis)

Specifications

  • Pack type:
    Glass bottle
  • Size:
    250 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.