Phenylsilane 97+%
Glass bottle, 5 ml
Catalog # TS29169-0050
Supplier: Thermo Scientific Chemicals
: AC29169-0050
Jump to:
Product Details & Documents
Synonyms: Silylbenzene
- CAS Number:694-53-1
- Formula:C₆H₈Si
- MDL Number:MFCD00013478
- Molecular Weight:108.22 g/mol
- Boiling Pt:120 °C (760 mmHg)
- Density:0.87 g/cm³
- Flash Pt:7 °C
- Storage Temperature:Ambient
- EINECS:211-772-5
- UN:3399
- ADR:4.3,II
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5090 to 1.5120 (20°C, 589 nm)
- GC:≥97.0 %
Specifications
- Pack type:Glass bottle
- Size:5 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



