Agar, Bacteriological Grade
Plastic bottle, 250 g
Catalog # TS44357-2500
Supplier: Thermo Scientific Chemicals
Supplier #: 44357-2500
Agar, Bacteriological Grade
Catalog # TS44357-2500
Supplier: Thermo Scientific Chemicals
CAS Number: 9002-18-0
Jump to:
Product Details & Documents
Synonyms: Agar Agar
- CAS Number:9002-18-0
- MDL Number:MFCD00081288
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:White to cream to light yellow to beige Powder
- Infrared spectrum:Conforms
- Loss on drying:≤15 % (1 g, 105°C)
- Solubility:(1.5 % in hot water) Clear to slightly opalescent light yellow to yellow solution
- Gel strength:≥700 g/cm² (1.5 %)
- Gel point:32 to 38°C (1.5 %)
- Ash:≤5 %
- pH:6.0 to 7.5 (10 % suspension)
Specifications
- Pack type:Plastic bottle
- Size:250 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



